2-Chloro-5-(trifluoromethyl)pyridin-3-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-2-chloro-5-(trifluoromethyl)pyridine
A111467-25G
A111467-1G
A111467-5G
A111467-10G
103797-838EA
9
USD
103797-838
103823-658
77025-788
77310-458
77310-460
77570-650
2-Chloro-5-(trifluoromethyl)pyridin-3-amine 98%
2-Chloro-5-(trifluoromethyl)pyridin-3-amine
Caution: for Research Use Only
Formula:
C₆H₄ClF₃N₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD07375383 CAS Number: 72587-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...