(-)-Epigallocatechin gallate 99%

Supplier: Ambeed
Warning

Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate, EGCG

A110153-100MG A110153-250MG A110153-25G A110153-5G A110153-1G A110153-100G
103797-626EA 27 USD
103797-626 77135-972 77135-974 77135-976 77135-978 77135-980 77283-952 77570-572
(-)-Epigallocatechin gallate 99%
(-)-Epigallocatechin gallate

Caution: for Research Use Only

Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
Boiling Pt: 909.1 °C (760 mm Hg)
Melting Pt: 216…218 °C
Flash Pt: 320 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075940
CAS Number: 989-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR