2-Bromo-4,5-difluorophenol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Bromo-3,4-difluoro-6-hydroxybenzene
A109988-1G
A109988-5G
A109988-25G
77283-916EA
9
USD
77283-916
77283-918
77570-560
2-Bromo-4,5-difluorophenol 98%
2-Bromo-4,5-difluorophenol
Caution: for Research Use Only
Formula:
C₆H₃BrF₂O |
MDL Number:
MFCD00070751 CAS Number: 166281-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...