IVD-US Spot Saver Card
Supplier: REVVITY HEALTH SCIENCES, INC.
GRNK11022
77569-990CS 852.57 USD
77569-990
IVD-US Spot Saver Card
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
GRNK1 cards are a medium for the collection and transportation of blood specimen spots to a laboratory, incorporating the high quality Ahlstrom 226 grade paper which ensures even and uniform sample distribution.
  • Two part card
  • 226 filter paper, 125# white tag and flap to cover blood spots
  • Code 3 of 9 Barcode with one human readable (serial number)
  • Blood collection circles Int.Ø = 12.7 mm
  • Final folded card size: 3²/₃(W) x 2⁵/₈"(H)
For in vitro diagnostic use.