Copper(II) oxide 99%

Supplier: Ambeed
Danger

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated

A989426-25G A989426-500G A989426-1000G A989426-100G
76698-948EA 11 USD
76698-948 77569-716 77603-402 77880-803
Copper(II) oxide 99%
Copper (II) oxide

Caution: For research use only.

Formula: CuO
MDL Number: MFCD00010979
CAS Number: 1317-38-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR