n-Undecane 99%

Supplier: Ambeed
Danger

Synonyms: Undecane

A538321-25G
77569-048EA 6.18 USD
77569-048
n-Undecane 99%
n-Undecane

Caution: For research use only.

Formula: H₃C(CH₂)₉CH₃
MW: 156.31 g/mol
MDL Number: MFCD00008959
CAS Number: 1120-21-4
UN: 2330
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR