Copper(I) iodide 99%
Supplier: Ambeed
|
Danger
|
Synonyms:
Cuprous iodide
A531002-25G
A531002-5G
A531002-100G
A531002-500G
A531002-1000G
103839-676EA
8
USD
103839-676
77307-736
77307-738
77551-606
77551-608
77569-028
Copper(I) iodide 99%
Copper (I) iodide
Caution: For research use only.
Formula:
CuI Boiling Pt: 1290 °C (760 mm Hg) Melting Pt: 605 °C Flash Pt: 1290 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010978 CAS Number: 7681-65-4 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...