EDTA (ethylenediamine tetraacetic acid) 98%

Supplier: Ambeed
Warning

A287527-100G A287527-25G A287527-500G A287527-1000G
76802-326EA 9 USD
76802-326 76802-328 76802-330 77568-584
EDTA (ethylenediamine tetraacetic acid) 98%
EDTA (Ethylenediamine tetraacetic acid)

Caution: For research use only.

Formula: C₁₀H₁₆N₂O₈
MDL Number: MFCD00003541
CAS Number: 60-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR