Tristearin (glycerol tristearate) 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propane-1,2,3-triyl tristearate, Tristearin
A221844-1000G
A221844-100G
A221844-500G
77550-438EA
23
USD
77550-438
77568-138
77600-776
Tristearin (glycerol tristearate) 97%
Tristearin (Glycerol tristearate)
Caution: For research use only.
Formula:
C₅₇H₁₁₀O₆ |
MDL Number:
MFCD00036230 CAS Number: 555-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...