Chloro(1,5-cyclooctadiene)iridium(I) dimer 97%

Supplier: Ambeed
Warning

Synonyms: [Ir(cod)Cl]2, Chloro-1,5-cyclooctadiene iridium(I) dimer

A132084-1G A132084-250MG A132084-5G A132084-100MG
76688-672EA 199 USD
76688-672 76688-674 76688-676 77566-562
Chloro(1,5-cyclooctadiene)iridium(I) dimer 97%
Chloro(1,5-cyclooctadiene)iridium (I) dimer

Caution: For research use only.

Formula: C₁₆H₂₄Cl₂Ir₂
MW: 671.70 g/mol
MDL Number: MFCD00012414
CAS Number: 12112-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR