2-Hydroxy-n-octanoic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Hydroxy-n-octanoic Acid, 2-Hydroxyoctanoic acid
A1194709-5G
A1194709-1G
A1194709-25G
A1194709-100G
77566-204EA
57
USD
77566-204
77571-160
77620-740
77866-510
2-Hydroxy-n-octanoic acid 95%
2-Hydroxy-n-octanoic acid
Caution: For research use only.
Formula:
C₈H₁₆O₃ MW: 160.21 g/mol |
MDL Number:
MFCD00014410 CAS Number: 617-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...