PI(4)P:PS Lipid Substrates
Supplier: Sino Biological
P427-59-1000 P427-59-500
77561-812EA 226 USD
77561-812 77561-814
PI(4)P:PS Lipid Substrates
Assays Protein Assays
The substrate solution used for assaying lipid kinases.
Formulation: Frozen substrate solution containing 0.25 mM of Phospha-tidylinositol 4-phosphate [PI(4)P or PtdIns4P] and 2 mM of Phosphatidylserine (PS) in 1X lipid dilution buffer
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.