PI(3)P Lipid Substrates
Supplier: Sino Biological
P421-59-05 P421-59-1000
77561-788EA 270 USD
77561-788 77561-790
PI(3)P Lipid Substrates
Assays Protein Assays
The substrate solution used for assaying lipid kinases.
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Formulation: Frozen substrate solution containing 1 mM of Phospha-tidylinositol 3-phosphate [PI(3)P or PtdIns3P] in 1X lipid dilution buffer