PI(3)P Lipid Substrates
Supplier: Sino Biological
The substrate solution used for assaying lipid kinases.
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Formulation: Frozen substrate solution containing 1 mM of Phospha-tidylinositol 3-phosphate [PI(3)P or PtdIns3P] in 1X lipid dilution buffer