Phosphatase Assay Buffers I
Supplier: Sino Biological
P01-09-05
P01-09-20
P01-09-01
77560-888EA
61
USD
77560-888
77560-890
77560-886
Phosphatase Assay Buffers I
Buffers
Assay Buffers
Stock solution of buffer used for assaying phosphatase protein.
- Frozen solution of buffer containing: 125 mM HEPES pH 7.2, 250 mM NaCl, 12.5 mM EDTA
- Buffer passed through a 0.2 µ filter membrane and packaged aseptically
- Store at −20 °C
For optimal storage, aliquot buffer into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...