Methyltransferase Dilution Buffers III
Supplier: Sino Biological
Solution of (1X) buffer used for diluting enzyme DNA methyltransferases.
Storage: For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.