SymLine ARNOLD Waste Funnel Bundles, V2.0
Supplier: SCAT AMERICAS, INC.
75352 75353
77553-694EA 385.96 USD
77553-694 77553-696
SymLine ARNOLD Waste Funnel Bundles, V2.0
Funnels Safety Funnels
Waste funnel bundle with 1x safety funnel 'ARNOLD', V2.0.
Electrostatic conductive plastic: Yes
Grounding connection: Yes
Hinged lid: Yes
Main material: PE-HD-EL
Thread size: S60/61
Certifications: UN-Y approved.