Dough Cutter/Scrapers
Supplier: Remco Products
40572 40573 40574 40575 40576 40577
77553-348EA 16.34 USD
77553-348 77553-350 77553-344 77553-352 77553-354 77553-346
Dough Cutter/Scrapers
Scrapers
The dough cutter/scraper cuts and handles dough, removes sticky messes that would clog a brush, and easily deals with burnt-on foods on tables and cutting boards.
  • Baker’s best companion
  • Blade is slightly rounded on the edges to avoid cutting users hands
  • Polypropylene grip is durable and moulded securely around the blade
  • Ergonomic finger rest makes the dough cutter/scraper easy to grip and use repetitively
  • Drop-shaped hanging hole makes storage on a wall bracket or shadow board easy
The flexible, 0.5 mm stainless-steel blade is excellent at cutting many types of dough, cooked pastries, and other soft foods. This scraper is an essential tool for the food industry, including processing plants, commercial kitchens, restaurants, and supermarkets.
Blade thickness: 0.5"
Min. usage temperature: −20 °C; Max usage temperature: 170 °C
Certifications: Complies with UK 2019 No. 704 on food contact materials, Meets the REACH Regulation (EC) No. 1907/2006, Complies with Halal and Kosher, Complies with (EC) 1935/2004 on food contact materials, Complies with FDA Regulationl CFR 21, Complies with EU Regulation 2023/2006/EC of Good Manufacturing Practice.