VWR® Stackable Shaking Incubator with Refrigeration
Supplier: VWR International
77553-334
77553-334EA 14650 USD
77553-334
VWR® Stackable Shaking Incubator with Refrigeration
Shakers and Mixers Shaking Incubators
A stackable 20 L incubated shaker with refrigeration and non-slip rubber mat.
  • Stainless steel chamber for easy cleaning
  • Digital controls
  • Magnetic clamping system for clamps/tubes/microplates
  • Adjustable static shelf
The VWR® stackable shaking incubators provide a space saving design that offers high-throughput capacity. With precision temperature control and high-grade stainless steel chambers, these shaking incubators provide industry-leading performance with a temperature accuracy of ±0.1 °C and a chamber uniformity of ±0.5 °C. Two models are available with temperature control up to 60 °C. A refrigerated model includes a powerful compressor, capable of maintaining sub-ambient temperatures as low as 4 °C.
The VWR® stackable shaking incubator series features an increased chamber height of nearly 23". This increased height provides compatibility with high-capacity Erlenmeyer flasks, accepting up to four 5 L Erlenmeyer flasks. An adjustable shelf is included with the incubator, supporting still incubations of plates and dishes, even while the shaking platform is in use.
Delivery information: Each incubator is supplied with a standard, non-skid silicone mat that is suitable for low-speed shaking applications. An optional, universal platform is also available for securing a wide range of flasks, tubes and other common vessels.