Nickel(II) bromide ≥96%

Supplier: Ambeed
Danger

Synonyms: Nickel dibromide

A835156-25G
77552-560EA 9 USD
77552-560
Nickel(II) bromide ≥96%
Nickel (II) bromide

Caution: for Research Use Only

Formula: Br₂Ni
MDL Number: MFCD00011141
CAS Number: 13462-88-9
UN: 3288
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR