Glyoxal 40% (w/w) in water

Supplier: Ambeed
Danger

A716938-100G
77552-194EA 6 USD
77552-194
Glyoxal 40% (w/w) in water
Glyoxal

Caution: for Research Use Only

Formula: C₂H₂O₂
MDL Number: MFCD00006957
CAS Number: 107-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR