Methyl chlorodifluoroacetate ≥98%

Supplier: Ambeed
Danger

A713051-25G
77552-186EA 24 USD
77552-186
Methyl chlorodifluoroacetate ≥98%
Methyl chlorodifluoroacetate

Caution: for Research Use Only

Formula: C₃H₃ClF₂O₂
MW: 144.51 g/mol
MDL Number: MFCD00000775
CAS Number: 1514-87-0
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR