(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥97%

Supplier: Ambeed
Danger

A643553-1G
77551-950EA 390 USD
77551-950
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥97%
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

Caution: for Research Use Only

Formula: C₁₀H₈ClF₃O₂
MDL Number: MFCD00044400
CAS Number: 39637-99-5
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR