1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane 98%

Supplier: Ambeed
Warning

Synonyms: (+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane, 1,2-Bis((2S,5S)-2,5-diphenylphospholan-1-yl)ethane, (S,S)-Ph-BPE

A626988-50MG A626988-1G A626988-250MG A626988-5G A626988-100MG
77145-128EA 62.83 USD
77145-128 77551-882 77551-884 77551-886 77573-034
1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane 98%
1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane

Caution: for Research Use Only

Formula: C₃₄H₃₆P₂
MDL Number: MFCD07369028
CAS Number: 824395-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR