1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane, 1,2-Bis((2S,5S)-2,5-diphenylphospholan-1-yl)ethane, (S,S)-Ph-BPE
A626988-50MG
A626988-1G
A626988-250MG
A626988-5G
A626988-100MG
77145-128EA
62.83
USD
77145-128
77551-882
77551-884
77551-886
77573-034
1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane 98%
1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane
Caution: for Research Use Only
Formula:
C₃₄H₃₆P₂ |
MDL Number:
MFCD07369028 CAS Number: 824395-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...