Triethylsilane ≥98%

Supplier: Ambeed
Danger

A334418-25G
77550-956EA 13 USD
77550-956
Triethylsilane ≥98%
Triethylsilane

Caution: for Research Use Only

Formula: C₆H₁₆Si
MDL Number: MFCD00009018
CAS Number: 617-86-7
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR