Palladium(II) acetate 98%

Supplier: Ambeed
Danger

Synonyms: Palladium diacetate, Palladium (II) acetate

A326671-250MG A326671-1G A326671-25G A326671-5G A326671-10G A326671-100MG
103825-444EA 62 USD
103825-444 103836-392 76724-058 76839-352 76839-354 76839-356 77550-904 77668-462
Palladium(II) acetate 98%
Palladium (II) acetate

Caution: for Research Use Only

Formula: C₄H₆O₄Pd
Melting Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00012453
CAS Number: 3375-31-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR