Isopropyl myristate 98%

Supplier: Ambeed
Warning

Synonyms: Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester, Isopropyl Myristate, Isopropyl tetradecanoate, Tetradecylic acid isopropyl ester

A260428-100G A260428-500G A260428-1000G
76801-778EA 13 USD
76801-778 76801-780 77550-636
Isopropyl myristate 98%
Isopropyl myristate

Caution: for Research Use Only

Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 320
Flash Pt: 150
MDL Number: MFCD00008982
CAS Number: 110-27-0
Merck Index: 13,05235

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR