Tetrabutylammonium dihydrogen phosphate 98%

Supplier: Ambeed
Warning

Synonyms: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

A240724-5G A240724-100G A240724-25G A240724-500G A240724-1000G
77550-548EA 6 USD
77550-548 77568-336 77568-338 77600-940 77667-976
Tetrabutylammonium dihydrogen phosphate 98%
Tetrabutylammonium dihydrogen phosphate

Caution: For research use only.

Formula: C₁₆H₃₈NO₄P
MW: 339.46 g/mol
MDL Number: MFCD00064526
CAS Number: 5574-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR