Potassium-tert-pentyloxide ≥98%

Supplier: Ambeed
Danger

Synonyms: Potassium tert-amylate, Potassium 2-methyl-2-butoxide, Potassium tert-pentoxide

A201641-25G A201641-100G
77550-296EA 40 USD
77550-296 77879-257
Potassium-tert-pentyloxide ≥98%
Potassium-tert-pentyloxide

Caution: for Research Use Only

Formula: C₅H₁₁KO
MDL Number: MFCD00064808
CAS Number: 41233-93-6
UN: 2921
ADR: 8,4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR