Fmoc-D-Thr(tBu)-OH 97%

Supplier: Ambeed
Warning

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-D-threonine, Fmoc-D-Thr(tBu)-OH, (2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(tert-butoxy)butanoic acid, N-Fmoc-O-tert-butyl-D-threonine

A183224-10G A183224-250MG A183224-1G A183224-25G A183224-5G A183224-100G
103815-620EA 7 USD
103815-620 103846-182 76784-658 76784-660 77301-824 77301-826 77301-828 77550-174
Fmoc-D-Thr(tBu)-OH 97%
Fmoc-D-Thr(tBu)-OH

Caution: for Research Use Only

Formula: C₂₃H₂₇NO₅
MW: 397.471 g/mol
Boiling Pt: 581.7 °C (760 mm Hg)
Melting Pt: 130 °C
Flash Pt: 305.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00080292
CAS Number: 138797-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR