n-Dodecyl-β-D-maltoside 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
A112981-10G
A112981-250MG
A112981-5G
A112981-25G
A112981-1G
76718-186EA
176
USD
76718-186
76718-188
76718-190
77549-190
77570-714
n-Dodecyl-β-D-maltoside 97%
n-Dodecyl-β-D-maltoside
Caution: for Research Use Only
Formula:
C₂₄H₄₆O₁₁ |
MDL Number:
MFCD00043012 CAS Number: 69227-93-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...