Fmoc-L-Tyr(Me)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Tyr(Me)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-methyl-L-tyrosine, N-Fmoc-4-methoxy-L-phenylalanine, N-Fmoc-O-methyl-L-tyrosine, Fmoc-Phe(4-OMe)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methoxy-L-phenylalanine

A108968-250MG A108968-1G A108968-5G A108968-10G
103797-418EA 6 USD
103797-418 103823-206 76869-622 77310-038 77310-040 77549-100
Fmoc-L-Tyr(Me)-OH 95%
Fmoc-L-Tyr(Me)-OH

Caution: for Research Use Only

Formula: C₂₅H₂₃NO₅
MW: 417.47 g/mol
Boiling Pt: 647.8 °C (760 mm Hg)
Flash Pt: 345.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153368
CAS Number: 77128-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR