Rapid Response™ Fentanyl Test Cassette
Supplier: BTNX INC.
FYL-1CAP156-25
77548-848PK 96.06 USD
77548-848
Rapid Response™ Fentanyl Test Cassette
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Rapid Response™ Fentanyl Test Cassette is a competitive binding, lateral flow immunochromatographic assay for qualitative detection of fentanyl in human urine at the cut-off concentration of 1 ng/ml. This single drug test cassette can provide accurate and fast results in 5 minutes.
  • Sample: Urine
  • Format: Cassette
  • Rapid test for detecting Fentanyl in human urine
  • Read results in 5 minutes
  • Individually packaged test cassettes with pipette droppers
To perform the test, place the test cassette on a clean and level surface. Hold the dropper vertically and add 3 drops of urine to the specimen well (S) of the test cassette. Read results at 5 minutes. Interpretation of results are: 2 colored lines, one in Control Region (C) and another in Test Region (T), is negative, 1 line in the Control Region (C) and no line in the Test Region (T) is positive, no lines or no line in the Control Region (C) is invalid.
Certifications: The Rapid Response™ Fentanyl Test Cassette is CLIA Waived and FDA Cleared for Point-of-Care in vitro diagnostic use only.
Delivery information: Each kit contains individually packaged test cassettes, pipette droppers, and instructions.
Caution: Please refer to the instruction provided in the kit before use. Read the entire procedure carefully prior to testing. Do not use after the expiration date indicated on the package. Do not reuse tests.