Xylene S
Supplier: G-Biosciences
786-2130
77540-318EA 35.82 USD
77540-318
Xylene S
Stains Microscopy and staining reagents
Xylene S is recommended to use in Wheatley Trichrome Stain Kit.
  • Highly specific staining solution