Gram color staining kit
Supplier: G-Biosciences
786-2105
77540-268KT 132.87 USD
77540-268
Gram color staining kit
Gram color staining kit
Gram staining kit is used to differentiate gram-positive bacteria from gram-negative bacteria.
  • Quick staining kit
Packaging: Kit includes 250 ml each of Gram Crystal Violet, Gram Safranin , Gram Decolorizer, and Gram Iodine