Gram color staining kit
Supplier: G-Biosciences
786-2104
77540-266KT 157.59 USD
77540-266
Gram color staining kit
Gram color staining kit
Gram staining kit, Advanced is used to differentiate gram-positive bacteria from gram-negative bacteria.
  • Quick staining kit
This kit includes 250 ml each of:
Gram crystal violet, advanced
Gram Safranin, advanced
Gram decolorizer, advanced
Gram iodine, advanced and
Gram enhancer