Crystal violet Advanced for Gram staining

Supplier: G-Biosciences

786-2083 786-2084
77540-224KT 153.47 USD
77540-224 77540-226
Crystal violet Advanced for Gram staining
Crystal Violet

Gram crystal violet is used for the differentiation of Gram-negative and Gram-positive microorganisms.

Formula: C₂₅H₃₀ClN₃
MDL Number: MFCD00011750
CAS Number: 548-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR