Crystal violet solution for Gram staining

Supplier: G-Biosciences

Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet

786-2079 786-2080 786-2081 786-2082
77540-216EA 296.51 USD
77540-216 77540-218 77540-220 77540-222
Crystal violet solution for Gram staining
Crystal Violet

A primary stain in the advanced Gram stain procedure.


  • An aqueous solution of crystal violet


Quick staining of Gram-positive and Gram-negative bacteria

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Number: MFCD00011750
CAS Number: 548-62-9
Merck Index: 13,04406

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR