Monarch® Spin Collection Tubes
Supplier: New England Biolabs (NEB)
T2118L
77532-262EA 26 USD
77532-262
Monarch® Spin Collection Tubes
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Monarch® Spin collection tubes are designed to be used with Monarch spin columns.
  • Compatible with all Monarch® spin columns
  • Designed and manufactured with less plastic than other leading suppliers
Monarch® Spin Collection Tubes are designed to be used with Monarch® Spin Columns for collection of waste and other fractions during centrifugation steps in Monarch® Spin Nucleic Acid Purification Kits.
Ordering information: Available for purchase separately. Included with Monarch Spin nucleic acid purification kits.
Packaging: Supplied as 100 collection tubes, in 2 packs of 50 tubes each.