Paper Labels for 3" Core Printers
Supplier: Brady Worldwide
30240
77532-232RL 42.57 USD
77532-232
Paper Labels for 3" Core Printers
Labels
General purpose paper with matte finish and smooth top coat finish for thermal transfer printing.
  • White matte paper label stock
  • Permanent rubber adhesive
  • Low cost is ideal for all general purpose labeling projects
Ideal for use in general labeling and barcoding applications where the label needs to be removed or repositionable. A ribbon is required to print this material.