Overhead Stirrer Multi Mixer, 1000
Supplier: GRANT USA INC.
MM1000
77531-722EA 897 USD
77531-722
Overhead Stirrer Multi Mixer, 1000
Stirrers Overhead Stirrers
The Multi mixer 1000 is designed for stirring liquids up to 20 litres.
  • Mixing, stirring and shearing in
  • Biotechnology
  • Organic synthesis
  • Analytical laboratories
  • Sample preparation
It is a quiet and reliable mixer that provides stable continuous mixing for up to seven days. It features three types of motion: Rotational, reciprocal and vibration.
It is an ideal instrument for biotechnology, organic synthesis, analytical laboratories. It can be used for stirring solutions up to the 'medium viscosity' range (from 1000 to 10000 mPa).
Ordering information: Five optional paddles, retort stand and clamp sold separately.