CO₂ Shaker, CPS - 20
Supplier: GRANT USA INC.
CPS20
77531-684EA 2088 USD
77531-684
CO₂ Shaker, CPS - 20
Shakers and Mixers Shaking Incubators
CO₂ shaker CPS - 20 provides regulated orbital motion of the platform and is designed for use specifically in CO₂ incubators.
  • Tissue culture experiments
  • Eukaryotic cell cultivations
  • Any orbital shaking application in CO₂ incubators
  • Brushless motor with a guaranteed life time of 35,000 hours
  • Digital time setting: 1 min to 96 hr or non-stop (Increment - one minute)
CPS - 20 is specifically designed for use in harsh environments such as CO₂ and humidity and provides reproducible results for cell culture growth. A choice of five interchangeable platforms are available to provide a wide variety of vessels to be used. The specially designed remote controller allows for protection of electronics from the CO₂ environment. It does not interfere with the operation of the incubator.
Ordering information: Five interchangeable platforms available (must be ordered separately).