Bouffant Cap
Supplier: O&M HALYARD, INC
69803
77531-664CS 131.26 USD
77531-664
Bouffant Cap
Disposable Garments
Halyard X-Large, bouffant cap, with elastic closure.
  • Made with lightweight spunbond fabric and are designed to cover hair in no fluid contamination environments
  • Dimensions: 27" circumference
  • Disposable