S-Klenz® Scale Remover
Supplier: Steris
1J8108
77531-584CS 349.7 USD
77531-584
S-Klenz® Scale Remover
Cleaning Agents Detergents and Surface Cleansers
S-Klenz Scale Remover is a phosphoric acid-based film and scale remover. This instrument rust inhibitor is also used for de-scaling equipment.
  • Phosphoric acid-based de-scaler effectively removes scale deposits
  • Free-rinsing surgical instrument scale remover leaves no residue behind in automated washer/disinfectors
  • Easily measured and dispensed liquid formula
Effective at removing lime scale, urine scale, and corrosion from stainless steel, glass, and all plastics. The wetting and penetrating agents of the formula remove film and scale, but is not recommended for use on aluminum surfaces, galvanized metals, and mild steel.