Septihol® Ethyl WFI se Alcohol Solution

Supplier: Steris
625608WR
77531-088CS 295 USD
77531-088
Septihol® Ethyl WFI se Alcohol Solution
Cleaning Agents Disinfectants
Disinfectant for microbial control.

  • Efficacy - Biocidal and fungicidal EPA claims
  • Productivity - Ready to use alcohol eliminates in-house production validation and documentation concerns
  • Rinsing - Evaporates quickly with no residue
  • Reliability - Each lot is tested to ensure low endotoxins

Septihol® Ethyl WFI se Alcohol Solution provides microbial control and removes residue from hard surfaces. It can be used straight out of the container and be applied poured or sprayed. Since it contains no stabilizers or denaturants, it evaporates quickly and completely.

Septihol® Ethyl WFI se is a non sterile mixture of ethanol, isopropanol, methanol and United States Pharmacopeia (USP) Water For Injection (WFI) designed to remove organic and inorganic residues in aseptic processing areas. Its EPA biocidal and fungicidal claims are proven in presence of 5% organic soil on hard, nonporous surfaces. The formula does not contain stabilizers or denaturants, so it evaporates quickly and completely.

CAS: 64-17-5; 67-63-0; 67-56-1
UN 1170
Hazard class: 3
Packing group: II

Caution: Danger, GHS02; GHS07; GHS08.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR