Sol-M® Blood Culture Tube Holder
Supplier: SOL-MILLENNIUM MEDICAL
110201050002
77528-888CS 154.46 USD
77528-888
Sol-M® Blood Culture Tube Holder
Blood Collection Tube Holders
The Sol-M® blood culture tube holder fits all standard evacuated blood culture bottles.
Threads are compatible with standard blood collection needles.
Packaging: Sterile and individually wrapped.