Human GP6 (Glycoprotein VI, Platelet) ELISA Kit

Supplier: AFG BIOSCIENCE LLC

EK242174
77514-764EA 385.7 USD
77514-764
Human GP6 (Glycoprotein VI, Platelet) ELISA Kit
Assays ELISAs
New Product

Order Now

SPECIFICATIONS

Target protein GP6 (Glycoprotein VI, Platelet)
Environmentally Preferable
Primary antibody reactivity
Assay Type Sandwich
Host Human
Sensitivity 6.3 pg/ml
Detection Range 15.63 - 1000 pg/ml
Sample Type Plasma
Time to Results 3.5 h
Regulatory Status RUO
Storage Temperature 2…8 °C
Shelf Life 6 months (4 °C); 12 months (–20 °C)
Application Metabolic pathway, hematology, cardiovascular biology

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR