Yeast Extract, Granulated
Supplier: RPI
Y20025-1000.0 Y20025-2000.0 Y20025-500.0 Y20025-5000.0 Y20025-10000.0
76348-884EA 219.31 USD
76348-884 76348-886 76348-888 76348-890 77510-510
Yeast Extract, Granulated
Media for Microbiology Dehydrated Media for Microbiology
Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins. Material is specially granulated to reduce dust and increase solubility rate. Suitable for Microbiology applications.
Caution: Research or further manufacturing use only, not for food or drug use.