Water, meets ASTM Type II specifications

Supplier: RPI

W35100-10000.0
77510-508EA 94.54 USD
77510-508
Water, meets ASTM Type II specifications
Water

DI water (ASTM TYPE II).


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Appearance Clear Liquid
Resistivity > 1 MΩ-cm
Conductivity <1 µS/cm
Total Organic Carbon (TOC) <50 ppb
Sodium <5 ppb
Chloride <5 ppb
Silica <3 ppb
Country of Origin US


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR